missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzaldehyde, 97%

Catalog No. p-7021121
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1332503 1 g
AAA1332506 5 g
AAA1332514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1332503 Supplier Thermo Scientific Chemicals Supplier No. A1332503
Only null left

CAS: 10203-08-4 | C7H4Cl2O | 175.008 g/mol

3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10203-08-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003352
InChI Key CASRSOJWLARCRX-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dichloro, 3,5-dichloro-benzaldehyde, unii-m74yu7se79, rarechem aq a2 0002, pubchem3068, 3,5-dichlorobezaldehyde, 3,5-dichlorobenaldehyde, 3?5-dichlorobenzaldehyde, benzaldehyde,5-dichloro, acmc-1bxq6
PubChem CID 35746
IUPAC Name 3,5-dichlorobenzaldehyde
SMILES C1=C(C=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 60°C to 66°C
Boiling Point 235°C to 240°C
Quantity 1 g
UN Number UN1759
Beilstein 2040553
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzaldehyde
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 233-499-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only