missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzaldehyde, 97%
Description
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 60°C to 66°C |
| Boiling Point | 235°C to 240°C |
| Quantity | 1 g |
| UN Number | UN1759 |
| Beilstein | 2040553 |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 175.01 |
| Percent Purity | 97% |
| Chemical Name or Material | 3,5-Dichlorobenzaldehyde |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S. |
| EINECSNumber | 233-499-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only