missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroaniline 98.0+%, TCI America™

Catalog No. D032525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032525G 25 g
D0325100G 100 g
D0325500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D032525G Supplier TCI America Supplier No. D032525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-43-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007774
InChI Key UQRLKWGPEVNVHT-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
PubChem CID 12281
ChEBI CHEBI:19904
IUPAC Name 3,5-dichloroaniline
SMILES C1=C(C=C(C=C1Cl)Cl)N

Specifications

Melting Point 51°C
Boiling Point 260°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloroaniline
EINECSNumber (3)-0261
TSCA Yes