missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5'-Dichloroacetophenone 98.0+%, TCI America™
$72.70 - $197.59
Chemical Identifiers
| CAS | 14401-72-0 |
|---|---|
| Molecular Formula | C8H6Cl2O |
| Molecular Weight (g/mol) | 189.035 |
| MDL Number | MFCD00045189 |
| InChI Key | JGMBBKVZFUHCJC-UHFFFAOYSA-N |
| PubChem CID | 2758058 |
| IUPAC Name | 1-(3,5-dichlorophenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC(=C1)Cl)Cl |
Chemical Identifiers
| 14401-72-0 | |
| 189.035 | |
| JGMBBKVZFUHCJC-UHFFFAOYSA-N | |
| 1-(3,5-dichlorophenyl)ethanone |
| C8H6Cl2O | |
| MFCD00045189 | |
| 2758058 | |
| CC(=O)C1=CC(=CC(=C1)Cl)Cl |
Specifications
| 14401-72-0 | |
| 135°C | |
| MFCD00045189 | |
| JGMBBKVZFUHCJC-UHFFFAOYSA-N | |
| 1-(3,5-dichlorophenyl)ethanone | |
| 2758058 | |
| ≥98.0% (GC) | |
| 3′,5′-Dichloroacetophenone |
| Yellow | |
| C8H6Cl2O | |
| 1 g | |
| CC(=O)C1=CC(=CC(=C1)Cl)Cl | |
| 189.035 | |
| 189.04 | |
| Liquid |
Safety and Handling
TSCA : No