missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-iodopyridine 98.0+%, TCI America™

Catalog No. D51955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51955G 5 g
D51951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51955G Supplier TCI America Supplier No. D51955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 343781-41-9 | C5H2Cl2IN | 273.88 g/mol

Chemical Identifiers

CAS 343781-41-9
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.88
MDL Number MFCD03094687
InChI Key DEXLTMBFLWCGNF-UHFFFAOYSA-N
PubChem CID 2736004
IUPAC Name 3,5-dichloro-4-iodopyridine
SMILES ClC1=CN=CC(Cl)=C1I

Specifications

Melting Point 189°C
Quantity 5 g
Formula Weight 273.88
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-4-iodopyridine