missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5'-Dichloro-4'-hydroxyacetophenone 98.0+%, TCI America™
$240.46 - $710.21
Chemical Identifiers
| CAS | 17044-70-1 |
|---|---|
| Molecular Formula | C8H6Cl2O2 |
| Molecular Weight (g/mol) | 205.034 |
| MDL Number | MFCD00016421 |
| InChI Key | FXSIZYWHUQEXPC-UHFFFAOYSA-N |
| PubChem CID | 86920 |
| IUPAC Name | 1-(3,5-dichloro-4-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=C(C(=C1)Cl)O)Cl |
Chemical Identifiers
| 17044-70-1 | |
| 205.034 | |
| FXSIZYWHUQEXPC-UHFFFAOYSA-N | |
| 1-(3,5-dichloro-4-hydroxyphenyl)ethanone |
| C8H6Cl2O2 | |
| MFCD00016421 | |
| 86920 | |
| CC(=O)C1=CC(=C(C(=C1)Cl)O)Cl |
Specifications
| 17044-70-1 | |
| C8H6Cl2O2 | |
| 1 g | |
| CC(=O)C1=CC(=C(C(=C1)Cl)O)Cl | |
| 205.034 | |
| 205.03 | |
| Crystalline Powder |
| 164°C | |
| MFCD00016421 | |
| FXSIZYWHUQEXPC-UHFFFAOYSA-N | |
| 1-(3,5-dichloro-4-hydroxyphenyl)ethanone | |
| 86920 | |
| ≥98.0% (GC,T) | |
| 3′,5′-Dichloro-4′-hydroxyacetophenone |
Safety and Handling
TSCA : No