missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5'-Dichloro-2'-hydroxyacetophenone 98.0+%, TCI America™
Supplier: TCI America D474225G
Specifications
| 3′,5′-Dichloro-2′-hydroxyacetophenone | |
| 97°C | |
| 134°C | |
| MFCD00016417 | |
| 1-3,5-dichloro-2-hydroxyphenyl ethanone, 3',5'-dichloro-2'-hydroxyacetophenone, 3,5-dichloro-2-hydroxyacetophenone, 1-3,5-dichloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4,6-dichlorophenol, 2'-hydroxy-3',5'-dichloroacetophenone, ethanone, 1-3,5-dichloro-2-hydroxyphenyl, 1-acetyl-3,5-dichloro-2-hydroxybenzene, acmc-1cq4k, 4,6-dichloro-2-acetylphenol | |
| CC(=O)C1=CC(=CC(=C1O)Cl)Cl | |
| 205.034 | |
| 205.03 | |
| Crystalline Powder |
| 3321-92-4 | |
| White-Yellow | |
| C8H6Cl2O2 | |
| 25 g | |
| CJFYGRLJDKWMDI-UHFFFAOYSA-N | |
| 1-(3,5-dichloro-2-hydroxyphenyl)ethanone | |
| 520608 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3321-92-4 | |
| 205.034 | |
| CJFYGRLJDKWMDI-UHFFFAOYSA-N | |
| 520608 | |
| CC(=O)C1=CC(=CC(=C1O)Cl)Cl |
| C8H6Cl2O2 | |
| MFCD00016417 | |
| 1-3,5-dichloro-2-hydroxyphenyl ethanone, 3',5'-dichloro-2'-hydroxyacetophenone, 3,5-dichloro-2-hydroxyacetophenone, 1-3,5-dichloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4,6-dichlorophenol, 2'-hydroxy-3',5'-dichloroacetophenone, ethanone, 1-3,5-dichloro-2-hydroxyphenyl, 1-acetyl-3,5-dichloro-2-hydroxybenzene, acmc-1cq4k, 4,6-dichloro-2-acetylphenol | |
| 1-(3,5-dichloro-2-hydroxyphenyl)ethanone |
Safety and Handling
TSCA : No