missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dichloro-2'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. D47425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47425G 5 g
D474225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47425G Supplier TCI America Supplier No. D47425G

Chemical Identifiers

CAS 3321-92-4
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00016417
InChI Key CJFYGRLJDKWMDI-UHFFFAOYSA-N
Synonym 1-3,5-dichloro-2-hydroxyphenyl ethanone, 3',5'-dichloro-2'-hydroxyacetophenone, 3,5-dichloro-2-hydroxyacetophenone, 1-3,5-dichloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4,6-dichlorophenol, 2'-hydroxy-3',5'-dichloroacetophenone, ethanone, 1-3,5-dichloro-2-hydroxyphenyl, 1-acetyl-3,5-dichloro-2-hydroxybenzene, acmc-1cq4k, 4,6-dichloro-2-acetylphenol
PubChem CID 520608
IUPAC Name 1-(3,5-dichloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dichloro-2′-hydroxyacetophenone