missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dichloro-2'-hydroxyacetophenone, 99%

Catalog No. p-7045258
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1000606 5 g
AAL1000614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1000606 Supplier Thermo Scientific Chemicals Supplier No. L1000606
Only null left

CAS: 3321-92-4 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3321-92-4
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00016417
InChI Key CJFYGRLJDKWMDI-UHFFFAOYSA-N
Synonym 1-3,5-dichloro-2-hydroxyphenyl ethanone, 3',5'-dichloro-2'-hydroxyacetophenone, 3,5-dichloro-2-hydroxyacetophenone, 1-3,5-dichloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4,6-dichlorophenol, 2'-hydroxy-3',5'-dichloroacetophenone, ethanone, 1-3,5-dichloro-2-hydroxyphenyl, 1-acetyl-3,5-dichloro-2-hydroxybenzene, acmc-1cq4k, 4,6-dichloro-2-acetylphenol
PubChem CID 520608
IUPAC Name 1-(3,5-dichloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 95°C to 98°C
Boiling Point 132°C to 134°C (18 mmHg)
Quantity 5 g
Beilstein 1638099
Formula Weight 205.04
Percent Purity 99%
Chemical Name or Material 3',5'-Dichloro-2'-hydroxyacetophenone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only