missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichloro-2,4,6-trifluoropyridine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 311400050
Specifications
| 3,5-Dichloro-2,4,6-trifluoropyridine | |
| 1.6220g/mL | |
| 113°C | |
| Glass bottle | |
| 5g | |
| 2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6 | |
| C1(=C(C(=NC(=C1Cl)F)F)Cl)F | |
| 201.96 | |
| 201.96 |
| 1737-93-5 | |
| 159.0°C to 160.0°C | |
| 97.5% min. (GC) | |
| C5Cl2F3N | |
| 1.622 | |
| PKSORSNCSXBXOT-UHFFFAOYSA-N | |
| 3,5-dichloro-2,4,6-trifluoropyridine | |
| 61273 | |
| 98% |
Chemical Identifiers
| 1737-93-5 | |
| 201.96 | |
| 2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6 | |
| 3,5-dichloro-2,4,6-trifluoropyridine |
| C5Cl2F3N | |
| PKSORSNCSXBXOT-UHFFFAOYSA-N | |
| 61273 | |
| C1(=C(C(=NC(=C1Cl)F)F)Cl)F |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 217-088-3
RUO â Research Use Only