missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7126077
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01885G 5 g
D018825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01885G Supplier TCI America Supplier No. D01885G
Only null left

Chemical Identifiers

CAS 90-59-5
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.915
MDL Number MFCD00003318
InChI Key JHZOXYGFQMROFJ-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
PubChem CID 7024
IUPAC Name 3,5-dibromo-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Br

Specifications

Melting Point 83°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 279.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromosalicylaldehyde
RTECSNumber CU5609000
TSCA No