missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylaldehyde, 98%

CAS: 90-59-5 | C7H4Br2O2 | 279.915 g/mol

Supplier:  Thermo Scientific Chemicals A1372918

 View more versions of this product

Catalog No. AAA1372918


Only null left
Explore available promotions

Description

Description

3,5-Dibromosalicylaldehyde reacts with alkyl cyanoacetates in the presence of ammonium acetate to yield 4H- chromenes. It can be used in the synthesis of Schiff base and can be used as reactant for synthesis of Schiff base ligands which forms mononuclear complexes with copper(II), nickel(II), zinc(II) and cobalt(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromosalicylaldehyde reacts with alkyl cyanoacetates in the presence of ammonium acetate to yield 4H- chromenes. It can be used in the synthesis of Schiff base and can be used as reactant for synthesis of Schiff base ligands which forms mononuclear complexes with copper(II), nickel(II), zinc(II) and cobalt(II).

Solubility
Soluble in methanol 25 mg/mL. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air Sensitive.
Specifications

Specifications

3,5-Dibromosalicylaldehyde
81°C to 84°C
MFCD00003318
1424739
14,3027
Soluble in methanol 25mg/mL. Insoluble in water.
C1=C(C=C(C(=C1Br)O)C=O)Br
279.915
279.93
90-59-5
C7H4Br2O2
50 g
Air Sensitive
3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
JHZOXYGFQMROFJ-UHFFFAOYSA-N
3,5-dibromo-2-hydroxybenzaldehyde
7024
98%

Chemical Identifiers

90-59-5
279.915
JHZOXYGFQMROFJ-UHFFFAOYSA-N
7024
C1=C(C=C(C(=C1Br)O)C=O)Br
C7H4Br2O2
MFCD00003318
3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
3,5-dibromo-2-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

EINECSNumber : 202-003-4

RTECSNumber : CU5609000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only