missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromopyridine N-Oxide 98.0+%, TCI America™
$109.98 - $365.35
Chemical Identifiers
| CAS | 2402-99-5 |
|---|---|
| Molecular Formula | C5H3Br2NO |
| Molecular Weight (g/mol) | 252.893 |
| MDL Number | MFCD00160750 |
| InChI Key | QBUQQUOGXPOBAQ-UHFFFAOYSA-N |
| PubChem CID | 620003 |
| IUPAC Name | 3,5-dibromo-1-oxidopyridin-1-ium |
| SMILES | C1=C(C=[N+](C=C1Br)[O-])Br |
Chemical Identifiers
| 2402-99-5 | |
| 252.893 | |
| QBUQQUOGXPOBAQ-UHFFFAOYSA-N | |
| 3,5-dibromo-1-oxidopyridin-1-ium |
| C5H3Br2NO | |
| MFCD00160750 | |
| 620003 | |
| C1=C(C=[N+](C=C1Br)[O-])Br |
Specifications
| 2402-99-5 | |
| White-Yellow | |
| MFCD00160750 | |
| 3,5-dibromopyridine 1-oxide, 3,5-dibromopyridine n-oxide, 3,5-dibromopyridine-n-oxide, 3,5-dibromopyridin-1-ium-1-olate, 3,5-dibromopyridine1-oxide, 3,5-dibromopyridine-1-oxide, pyridine, 3,5-dibromo-,1-oxide, pubchem6616, 3,5-dibromopyridin-1-ol, 3,5-dibromopyridine n oxide | |
| C1=C(C=[N+](C=C1Br)[O-])Br | |
| 252.893 | |
| 252.89 | |
| Crystalline Powder |
| 144°C | |
| C5H3Br2NO | |
| 1 g | |
| QBUQQUOGXPOBAQ-UHFFFAOYSA-N | |
| 3,5-dibromo-1-oxidopyridin-1-ium | |
| 620003 | |
| ≥98.0% (GC) | |
| 3,5-Dibromopyridine N-Oxide |
Safety and Handling
TSCA : No