missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine N-Oxide 98.0+%, TCI America™

Catalog No. D43265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43265G 5 g
D43261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43265G Supplier TCI America Supplier No. D43265G
Only null left

Chemical Identifiers

CAS 2402-99-5
Molecular Formula C5H3Br2NO
Molecular Weight (g/mol) 252.893
MDL Number MFCD00160750
InChI Key QBUQQUOGXPOBAQ-UHFFFAOYSA-N
PubChem CID 620003
IUPAC Name 3,5-dibromo-1-oxidopyridin-1-ium
SMILES C1=C(C=[N+](C=C1Br)[O-])Br

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
Synonym 3,5-dibromopyridine 1-oxide, 3,5-dibromopyridine n-oxide, 3,5-dibromopyridine-n-oxide, 3,5-dibromopyridin-1-ium-1-olate, 3,5-dibromopyridine1-oxide, 3,5-dibromopyridine-1-oxide, pyridine, 3,5-dibromo-,1-oxide, pubchem6616, 3,5-dibromopyridin-1-ol, 3,5-dibromopyridine n oxide
Formula Weight 252.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromopyridine N-Oxide