missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzyl bromide, 99%, Thermo Scientific™

Catalog No. p-3211801
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC379780050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC379780050 Supplier Thermo Scientific Chemicals Supplier No. 379780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 56908-88-4
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.83
MDL Number MFCD00052415
InChI Key PWTFRUXTAFBWBW-UHFFFAOYSA-N
Synonym 3,5-dibromobenzyl bromide, 1,3-dibromo-5-bromomethyl benzene, benzene, 1,3-dibromo-5-bromomethyl, alpha,3,5-tribromotoluene, acmc-20andg, 3,5-dibromobenzylbromide, 3,5-dibromo benzyl bromide, 1,3-bis bromanyl-5-bromomethyl benzene
PubChem CID 143427
IUPAC Name 1,3-dibromo-5-(bromomethyl)benzene
SMILES BrCC1=CC(Br)=CC(Br)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3CH2Br
Quantity 5g
Formula Weight 328.83
Percent Purity 99%
Chemical Name or Material 3, 5-Dibromobenzyl bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only