missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-hydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. D018925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D018925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D018925G Supplier TCI America Supplier No. D018925G
Only null left

Chemical Identifiers

CAS 2973-77-5
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016980
InChI Key SXRHGLQCOLNZPT-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dibromo-4-hydroxy, timtec-bb sbb006529, acmc-1cr6c, 3,5-dibromo-4-hydroxy benzaldehyde, 3,5-dibromo-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-dibromo benzaldehyde, 3,5-dibromo-4-hydroxybenzaldehyde, 3,5-bis bromanyl-4-oxidanyl-benzaldehyde
PubChem CID 18100
IUPAC Name 3,5-dibromo-4-hydroxybenzaldehyde
SMILES OC1=C(Br)C=C(C=O)C=C1Br

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 25 g
Formula Weight 279.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-4-hydroxybenzaldehyde