missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dibromo-2'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. D46375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46375G 5 g
D463725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46375G Supplier TCI America Supplier No. D46375G

Chemical Identifiers

CAS 22362-66-9
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.942
MDL Number MFCD00191992
InChI Key RCKHUOIKVIOMGZ-UHFFFAOYSA-N
Synonym 3',5'-dibromo-2'-hydroxyacetophenone, 1-3,5-dibromo-2-hydroxyphenyl ethanone, 1-3,5-dibromo-2-hydroxyphenyl ethan-1-one, 3´,5´-dibromo-2´-hydroxyacetophenone, 1-acetyl-3,5-dibromo-2-hydroxybenzene, acmc-1cp7q, 2-acetyl-4,6-dibromophenol, dibromooxochromenecarboxylicacid, 3/',5/'-dibromo-2/'-hydroxyacetophenone, ethanone,1-3,5-dibromo-2-hydroxyphenyl
PubChem CID 271762
IUPAC Name 1-(3,5-dibromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)Br)Br

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 293.94
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dibromo-2′-hydroxyacetophenone