missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dibromo-2'-hydroxyacetophenone, 99%

Catalog No. AAB2241306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2241306 5 g
AAB2241314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2241306 Supplier Thermo Scientific Chemicals Supplier No. B2241306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22362-66-9 | C8H6Br2O2 | 293.942 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22362-66-9
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.942
MDL Number MFCD00191992
InChI Key RCKHUOIKVIOMGZ-UHFFFAOYSA-N
Synonym 3',5'-dibromo-2'-hydroxyacetophenone, 1-3,5-dibromo-2-hydroxyphenyl ethanone, 1-3,5-dibromo-2-hydroxyphenyl ethan-1-one, 3´,5´-dibromo-2´-hydroxyacetophenone, 1-acetyl-3,5-dibromo-2-hydroxybenzene, acmc-1cp7q, 2-acetyl-4,6-dibromophenol, dibromooxochromenecarboxylicacid, 3/',5/'-dibromo-2/'-hydroxyacetophenone, ethanone,1-3,5-dibromo-2-hydroxyphenyl
PubChem CID 271762
IUPAC Name 1-(3,5-dibromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)Br)Br

Specifications

Melting Point 108°C to 111°C
Quantity 5 g
Formula Weight 293.95
Percent Purity 99%
Chemical Name or Material 3',5'-Dibromo-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only