missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-1H-1,2,4-triazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 7411-23-6
Molecular Formula C2HBr2N3
Molecular Weight (g/mol) 226.859
InChI Key FRAKFBWDPXYIQO-UHFFFAOYSA-N
Synonym 3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
PubChem CID 81904
IUPAC Name 3,5-dibromo-1H-1,2,4-triazole
SMILES C1(=NC(=NN1)Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MO07857DA
SDS View Documents
Thermo Scientific Chemicals
MO07857DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7411-23-6
226.859
3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
3,5-dibromo-1H-1,2,4-triazole
C2HBr2N3
FRAKFBWDPXYIQO-UHFFFAOYSA-N
81904
C1(=NC(=NN1)Br)Br

Specifications

7411-23-6
Amber Glass Bottle
1g
FRAKFBWDPXYIQO-UHFFFAOYSA-N
3,5-dibromo-1H-1,2,4-triazole
81904
97%
97%
C2HBr2N3
3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
C1(=NC(=NN1)Br)Br
226.859
226.86
3,5-Dibromo-1H-1,2,4-triazole
Videos
SDS
Documents

Documents