missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-1,2,4-triazole 97.0+%, TCI America™

Catalog No. p-7128620
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50305G 5 g
D503025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50305G Supplier TCI America Supplier No. D50305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7411-23-6
Molecular Formula C2HBr2N3
Molecular Weight (g/mol) 226.859
MDL Number MFCD00487260
InChI Key FRAKFBWDPXYIQO-UHFFFAOYSA-N
Synonym 3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
PubChem CID 81904
IUPAC Name 3,5-dibromo-1H-1,2,4-triazole
SMILES C1(=NC(=NN1)Br)Br

Specifications

Melting Point 212°C
Quantity 5 g
Formula Weight 226.86
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-1,2,4-triazole
RTECSNumber XZ4570000
TSCA No