missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibenzyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. D31841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31841G 1 g
D31845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31841G Supplier TCI America Supplier No. D31841G

Chemical Identifiers

CAS 14615-72-6
Molecular Formula C21H18O3
Molecular Weight (g/mol) 318.372
InChI Key CHUAMRVJSRBRHT-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzaldehyde
PubChem CID 561351
IUPAC Name 3,5-bis(phenylmethoxy)benzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)C=O)OCC3=CC=CC=C3

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 318.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzaldehyde