missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%

Catalog No. p-7080808
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1051409 10 g
AAA1051418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1051409 Supplier Thermo Scientific Chemicals Supplier No. A1051409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22014-01-3 | C17H24O3 | 276.38 g/mol

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.

Chemical Identifiers

CAS 22014-01-3
Molecular Formula C17H24O3
Molecular Weight (g/mol) 276.38
MDL Number MFCD00017291
InChI Key CTYWXRDQWMRIIM-BQYQJAHWSA-N
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl acrylic acid, unii-k7zj1m9j5t, k7zj1m9j5t, 3,5-di-tert-butyl-4-hydroxycinnamic acid, 3,5-di-t-butyl-4-hydroxycinnamic acid, 3,5-di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 2e-3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-bis tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-ditert-butyl-4-hydroxyphenyl acrylic acid
PubChem CID 689095
IUPAC Name (E)-3-(3,5-ditert-butyl-4-hydroxyphenyl)prop-2-enoic acid
SMILES CC(C)(C)C1=CC(\C=C\C(O)=O)=CC(=C1O)C(C)(C)C

Specifications

Melting Point ∼195°C (decomposition)
Quantity 10 g
Beilstein 3144594
Solubility Information Soluble in water.
Formula Weight 276.37
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only