missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. D158725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D158725G 25 g
D1587100G 100 g
D1587500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D158725G Supplier TCI America Supplier No. D158725G
Only null left

Chemical Identifiers

CAS 1620-98-0
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00008826
InChI Key DOZRDZLFLOODMB-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
PubChem CID 73219
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 234.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzaldehyde
RTECSNumber CU5610070
TSCA No