missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%
Description
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.
Solubility
Soluble in hot methanol.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.
Solubility
Soluble in hot methanol.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 187°C to 189°C |
| Quantity | 5 g |
| Beilstein | 982526 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in hot methanol. |
| Formula Weight | 234.34 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3,5-Di-tert-butyl-4-hydroxybenzaldehyde |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 216-592-0 |
| RTECSNumber | CU5610070 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only