missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%

Catalog No. p-7020681
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1272206 5 g
AAA1272214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1272206 Supplier Thermo Scientific Chemicals Supplier No. A1272206

CAS: 14035-33-7 | C16H24O2 | 248.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14035-33-7
Molecular Formula C16H24O2
Molecular Weight (g/mol) 248.37
MDL Number MFCD00017520
InChI Key WGJPGMJLARWHRK-UHFFFAOYSA-N
Synonym 1-3,5-di-tert-butyl-4-hydroxyphenyl ethanone, 1-3,5-ditert-butyl-4-hydroxyphenyl ethanone, 1-3,5-di tert-butyl-4-hydroxyphenyl ethan-1-one, 3',5'-di-tert-butyl-4'-hydroxyacetophenone, 3,5-di-tert-butyl-4-hydroxyacetophenone, 1-3,5-di-tert-butyl-4-hydroxyphenyl ethan-1-one, ethanone, 1-3,5-bis 1,1-dimethylethyl-4-hydroxyphenyl, 1-3,5-bis 1,1-dimethylethyl-4-hydroxyphenyl ethanone, 1-acetyl-3,5-bis tert-butyl-4-hydroxybenzene, cbmicro_019190
PubChem CID 616296
IUPAC Name 1-(3,5-ditert-butyl-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 146°C to 147°C
Quantity 5 g
Formula Weight 248.37
Percent Purity 98%
Chemical Name or Material 3',5'-Di-tert-butyl-4'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only