missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-2-thienyl)-5-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog No. cc19710da
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC19710DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC19710DA Supplier Thermo Scientific Chemicals Supplier No. CC19710DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 180530-13-6
Molecular Formula C7H5BrN2OS
Molecular Weight (g/mol) 245.094
MDL Number MFCD03407331
InChI Key ZNWQLJYQXULFQB-UHFFFAOYSA-N
Synonym 3-5-bromo-2-thienyl-5-methyl-1,2,4-oxadiazole, 3-5-bromothiophen-2-yl-5-methyl-1,2,4-oxadiazole, 3-2-bromothien-5-yl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-5-bromo-2-thienyl-5-methyl, 5-bromo-2-5-methyl 1,2,4-oxadiazol-3-yl thiophene
PubChem CID 2776504
IUPAC Name 3-(5-bromothiophen-2-yl)-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC=C(S2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 245.1
Percent Purity 97%
Chemical Name or Material 3-(5-Bromo-2-thienyl)-5-methyl-1, 2, 4-oxadiazole