missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-2-methoxyphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAH3396106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3396106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3396106 Supplier Thermo Scientific Chemicals Supplier No. H3396106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82547-30-6 | Molecular Formula: C10H11BrO3 | Molecular Weight: 259.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82547-30-6
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.10
MDL Number MFCD09258907
InChI Key RCLIOLQUIYTUPY-UHFFFAOYSA-N
Synonym 3-5-bromo-2-methoxyphenyl propanoic acid, benzenepropanoic acid, 5-bromo-2-methoxy, 3-5-bromo-2-methoxyphenyl propionic acid, 3-5-bromo-2-methoxy-phenyl-propionic acid
PubChem CID 22685184
IUPAC Name 3-(5-bromo-2-methoxyphenyl)propanoic acid
SMILES COC1=C(CCC(O)=O)C=C(Br)C=C1

Specifications

Melting Point 114°C to 116°C
Quantity 5 g
Formula Weight 259.1
Percent Purity 96%
Chemical Name or Material 3-(5-Bromo-2-methoxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only