missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)pyridine, 97%

Catalog No. AAL19447MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19447MD 250 mg
AAL1944703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19447MD Supplier Thermo Scientific Chemicals Supplier No. L19447MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20857-47-0 | C7H3F6N | 215.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20857-47-0
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00128904
InChI Key RQEOYYWUVYZZLL-UHFFFAOYSA-N
PubChem CID 2778311
IUPAC Name 3,5-bis(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1C(F)(F)F)C(F)(F)F

Specifications

Melting Point 35°C to 38°C
Boiling Point 117°C to 118°C
Flash Point >100°C (212°F)
Quantity 250 mg
Beilstein 1684576
Synonym 3,5-bis trifluoromethyl pyridine, pyridine, 3,5-bis trifluoromethyl, pubchem7698, acmc-1bncl, 3,5-bis-trifluoromethylpyridine, pyridine,3,5-bis trifluoromethyl
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only