missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%

Catalog No. AAL1767306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1767306 5 g
AAL1767303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1767306 Supplier Thermo Scientific Chemicals Supplier No. L1767306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33558-36-0 | C10H9F6Si | 271.257 g/mol

3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 33558-36-0
Molecular Formula C10H9F6Si
Molecular Weight (g/mol) 271.257
MDL Number MFCD02093948
InChI Key HJQABLGRCWBCDT-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyl dimethylsilane, 3,5-bis trifluoromethyl phenyl-dimethyl-silicon, 3,5-bis-trifluoromethyl phenyldimethylsilane, 3,5-bis trifluoromethyl phenyl-dimethylsilicon, 3,5-bis trifluoromethyl phenyldimethylsilane
PubChem CID 11219490
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 40°C (1.5mmHg)
Quantity 5 g
Beilstein 2859742
Formula Weight 272.27
Percent Purity 95%
Refractive Index 1.4125
Density 1.23
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyldimethylsilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only