missing translation for 'onlineSavingsMsg'
Learn More

3',5,7-Trihydroxy-4'-methoxyflavanone, 97%

Catalog No. AAB2052803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2052803 1 g
AAB2052806 5 g
AAB2052814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2052803 Supplier Thermo Scientific Chemicals Supplier No. B2052803

CAS: 520-33-2 | C16H14O6 | 302.28 g/mol

(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 520-33-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00075646
InChI Key AIONOLUJZLIMTK-AWEZNQCLSA-N
Synonym hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1
PubChem CID 72281
ChEBI CHEBI:28230
IUPAC Name (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 227°C to 232°C
Quantity 1 g
Beilstein 309850
Merck Index 14,4670
Solubility Information Soluble in water (partly),dilute alkalis,and ethanol (50mg/ml).
Formula Weight 302.3
Percent Purity 97%
Chemical Name or Material 3',5,7-Trihydroxy-4'-methoxyflavanone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-290-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only