missing translation for 'onlineSavingsMsg'
Learn More

3,5,6-Trifluoro-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. T20615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20615G 5 g
T20611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20615G Supplier TCI America Supplier No. T20615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75777-49-0 | C5H2F3NO | 149.072 g/mol

Chemical Identifiers

CAS 75777-49-0
Molecular Formula C5H2F3NO
Molecular Weight (g/mol) 149.072
MDL Number MFCD04114299
InChI Key PWBGZZXJAMGVGQ-UHFFFAOYSA-N
Synonym 3,5,6-trifluoro-2-hydroxypyridine, 2-hydroxy-3,5,6-trifluoropyridine, 3,5,6-trifluoropyridin-2-ol, acmc-1beik, 3,5,6-trifluoro-2-pyridone, 3,5,6-trifluoro-2-pyridinol, 3,5,6-trifluoropyridin-2 1h-one, 2 1h-pyridinone,3,5,6-trifluoro, 3,5,6-trifluoro-1,2-dihydropyridine-2-one, 2 1h-pyridinone, 3,5,6-trifluoro-9ci
PubChem CID 23340297
IUPAC Name 3,5,6-trifluoro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC(=C1F)F)F

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 149.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5,6-Trifluoro-2-hydroxypyridine