missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5,6-Trifluoro-2-hydroxypyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 100°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 149.07 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,5,6-Trifluoro-2-hydroxypyridine |
Safety and Handling
| TSCA | No |