missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethyl-1-hexanol 85.0+%, TCI America™

Catalog No. T059625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T059625ML 25 mL
T0596500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T059625ML Supplier TCI America Supplier No. T059625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3452-97-9
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00036138
InChI Key BODRLKRKPXBDBN-UHFFFAOYNA-N
Synonym 3,5,5-Trimethylhexyl Alcohol
PubChem CID 18938
IUPAC Name 3,5,5-trimethylhexan-1-ol
SMILES CC(CCO)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 193°C
Quantity 25 mL
UN Number 3082
Formula Weight 144.26
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5,5-Trimethyl-1-hexanol
EINECSNumber (2)-0217
TSCA Yes