missing translation for 'onlineSavingsMsg'
Learn More

3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid, ≥97%, Thermo Scientific™

Catalog No. p-3110413
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30401CB 250mg
CC30401DA 1g
CC30401DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC30401CB Supplier Thermo Scientific Chemicals Supplier No. CC30401CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 500013-38-7
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.389
MDL Number MFCD04115219
InChI Key CXJBKOKDFABTQO-UHFFFAOYSA-N
Synonym 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoic acid, 1-3-carboxyphenylmethyl-4-boc piperazine, 1-boc-4-3-carboxy-benzyl-piperazine, 3-4-n-boc-piperazin-1-yl methylbenzoic acid, 1-tert-butoxycarbonyl-4-3-carboxybenzyl piperazine, tert-butyl 4-3-carboxybenzyl piperazine-1-carboxylate, 3-4-tert-butoxy carbonyl piperazin-1-yl methyl benzoic acid, 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoicacid, 1-piperazinecarboxylicacid, 4-3-carboxyphenyl methyl-, 1-1,1-dimethylethyl ester
PubChem CID 2795521
IUPAC Name 3-[[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]methyl]benzoic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 320.39
Percent Purity 97%
Chemical Name or Material 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.