missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)propionic acid, 97%

Catalog No. AAH5865503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5865503 1 g
AAH5865506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5865503 Supplier Thermo Scientific Chemicals Supplier No. H5865503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6318-43-0 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6318-43-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00084846
InChI Key WSXGQYDHJZKQQB-UHFFFAOYSA-N
Synonym 3-pyridin-4-yl propanoic acid, 3-pyridin-4-yl-propionic acid, 4-pyridinepropanoic acid, 4-pyridinepropionic acid, 3-4-pyridinyl propanoic acid, 3-4-pyridyl propanoic acid, 3-4-pyridyl propionic acid, 3-pyridin-4-yl propionic acid, 4-carboxyethylpyridine, acmc-1b6yz
PubChem CID 233243
IUPAC Name 3-pyridin-4-ylpropanoic acid
SMILES C1=CN=CC=C1CCC(=O)O

Specifications

Melting Point 231°C
Quantity 1 g
Beilstein 122234
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 3-(4-Pyridyl)propionic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only