missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-O-Isopropylidene-D-mannitol 96.0+%, TCI America™
$89.48 - $89.48
Chemical Identifiers
| CAS | 3969-84-4 |
|---|---|
| Molecular Formula | C9H18O6 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00075122 |
| InChI Key | YCOMFYACDCWMMD-UHFFFAOYNA-N |
| Synonym | 3,4-o-isopropylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol, 1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, oi4-o-isopropylidene-d-mannitol, d-mannitol-3,4-acetonide, 3-o,4-o-isopropylidene-d-mannitol, 3,4-isopropylidene-d-mannitol, 3,4-o-1-methylethylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol |
| PubChem CID | 7096260 |
| IUPAC Name | 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol |
| SMILES | CC1(C)OC(C(O)CO)C(O1)C(O)CO |
Chemical Identifiers
| 3969-84-4 | |
| 222.24 | |
| YCOMFYACDCWMMD-UHFFFAOYNA-N | |
| 7096260 | |
| CC1(C)OC(C(O)CO)C(O1)C(O)CO |
| C9H18O6 | |
| MFCD00075122 | |
| 3,4-o-isopropylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol, 1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, oi4-o-isopropylidene-d-mannitol, d-mannitol-3,4-acetonide, 3-o,4-o-isopropylidene-d-mannitol, 3,4-isopropylidene-d-mannitol, 3,4-o-1-methylethylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol | |
| 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol |
Specifications
| 3969-84-4 | |
| White-Yellow | |
| MFCD00075122 | |
| 3,4-o-isopropylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol, 1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, oi4-o-isopropylidene-d-mannitol, d-mannitol-3,4-acetonide, 3-o,4-o-isopropylidene-d-mannitol, 3,4-isopropylidene-d-mannitol, 3,4-o-1-methylethylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol | |
| CC1(C)OC(C(O)CO)C(O1)C(O)CO | |
| 222.24 | |
| 222.24 | |
| Crystalline Powder |
| 86°C | |
| C9H18O6 | |
| 1 g | |
| YCOMFYACDCWMMD-UHFFFAOYNA-N | |
| 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol | |
| 7096260 | |
| ≥96.0% (GC) | |
| 3,4-O-Isopropylidene-D-mannitol |
Safety and Handling
TSCA : No