missing translation for 'onlineSavingsMsg'
Learn More

3,4-O-Isopropylidene-D-mannitol 96.0+%, TCI America™

Catalog No. I04891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04891G Supplier TCI America Supplier No. I04891G
Only null left

Chemical Identifiers

CAS 3969-84-4
Molecular Formula C9H18O6
Molecular Weight (g/mol) 222.24
MDL Number MFCD00075122
InChI Key YCOMFYACDCWMMD-UHFFFAOYNA-N
Synonym 3,4-o-isopropylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol, 1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, oi4-o-isopropylidene-d-mannitol, d-mannitol-3,4-acetonide, 3-o,4-o-isopropylidene-d-mannitol, 3,4-isopropylidene-d-mannitol, 3,4-o-1-methylethylidene-d-mannitol, 1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol
PubChem CID 7096260
IUPAC Name 1-[5-(1,2-dihydroxyethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
SMILES CC1(C)OC(C(O)CO)C(O1)C(O)CO

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-O-Isopropylidene-D-mannitol