missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)propionic acid, 96%

Catalog No. p-7032266
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3366309 10 g
AAH3366318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3366309 Supplier Thermo Scientific Chemicals Supplier No. H3366309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16642-79-8 | C9H9NO4 | 195.17 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 16642-79-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00126834
InChI Key VZOPVJNBOQOLPN-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
PubChem CID 85526
IUPAC Name 3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 164°C to 168°C
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 195.17
Percent Purity 96%
Chemical Name or Material 3-(4-Nitrophenyl)propionic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only