missing translation for 'onlineSavingsMsg'
Learn More

3-(4-n-Propoxyphenyl)propionic acid, 96%

Catalog No. p-7032251
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3364603 1 g
AAH3364606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3364603 Supplier Thermo Scientific Chemicals Supplier No. H3364603

CAS: 3243-40-1 | C12H16O3 | 208.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3243-40-1
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00484121
InChI Key SOFMLHAGBATXNZ-UHFFFAOYSA-N
Synonym 3-4-propoxyphenyl propanoic acid, 3-4-propoxy-phenyl-propionic acid, benzenepropanoic acid,4-propoxy, 3-4-n-propoxyphenyl propionic acid, cbmicro_011397, cambridge id 5152838, 4-propoxy-benzenepropanoic acid, 3-4-n-propoxyphenyl propanoic acid
PubChem CID 2255935
IUPAC Name 3-(4-propoxyphenyl)propanoic acid
SMILES CCCOC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 94°C to 99°C
Quantity 1 g
Formula Weight 208.26
Percent Purity 96%
Chemical Name or Material 3-(4-n-Propoxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only