missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propylamine, 98%

Catalog No. p-7027040
Change view
Click to view available options
Quantity:
50 g
250 g
Catalog No. Quantity
AAB2348518 50 g
AAB2348530 250 g
2 options

Catalog No. AAB2348518

Supplier: Thermo Scientific Chemicals B2348518

Only null left

CAS: 123-00-2 | C7H16N2O | 144.22 g/mol

3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 123-00-2
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD00006184
InChI Key UIKUBYKUYUSRSM-UHFFFAOYSA-N
Synonym n-3-aminopropyl morpholine, 4-morpholinepropanamine, 3-morpholinopropylamine, 3-morpholinopropan-1-amine, n-aminopropylmorpholine, 4-3-aminopropyl morpholine, 4-morpholinepropylamine, 4-aminopropylmorpholine, 1-amino-3-morpholinopropane, 3-4-morpholinyl-1-propanamine
PubChem CID 61055
ChEBI CHEBI:86554
IUPAC Name 3-morpholin-4-ylpropan-1-amine
SMILES NCCCN1CCOCC1

Specifications

Melting Point -15°C
Density 0.987
Boiling Point 223°C to 225°C
Flash Point 98°C (210°F)
Refractive Index 1.4761
Quantity 50 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)propylamine
Show More Show Less
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H311-H314-H318
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 204-590-2
RTECSNumber QD7700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only