Learn More
3-(4-Morpholinyl)-1-propanol, 95%
CAS: 4441-30-9 | C7H15NO2 | 145.202 g/mol
$78.92 - $273.62
Chemical Identifiers
| CAS | 4441-30-9 |
|---|---|
| Molecular Formula | C7H15NO2 |
| Molecular Weight (g/mol) | 145.202 |
| MDL Number | MFCD01713533 |
| InChI Key | VZKSLWJLGAGPIU-UHFFFAOYSA-N |
| Synonym | 4-3-hydroxypropyl morpholine, 3-morpholinopropanol, 4-morpholinepropanol, 3-morpholin-4-yl propan-1-ol, 3-morpholinopropan-1-ol, 3-4-morpholinyl-1-propanol, 3-morpholin-4-yl-propan-1-ol, 3-morpholino-1-propanol, gamma-morpholinopropanol german, n-hydroxypropanylmorpholine |
| PubChem CID | 199576 |
| IUPAC Name | 3-morpholin-4-ylpropan-1-ol |
| SMILES | C1COCCN1CCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6037603
|
Thermo Scientific Chemicals
H6037603 |
1 g |
Each for $78.92
|
|
|||||
|
AAH6037606
|
Thermo Scientific Chemicals
H6037606 |
5 g |
Each for $273.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4441-30-9 | |
| 145.202 | |
| VZKSLWJLGAGPIU-UHFFFAOYSA-N | |
| 199576 | |
| C1COCCN1CCCO |
| C7H15NO2 | |
| MFCD01713533 | |
| 4-3-hydroxypropyl morpholine, 3-morpholinopropanol, 4-morpholinepropanol, 3-morpholin-4-yl propan-1-ol, 3-morpholinopropan-1-ol, 3-4-morpholinyl-1-propanol, 3-morpholin-4-yl-propan-1-ol, 3-morpholino-1-propanol, gamma-morpholinopropanol german, n-hydroxypropanylmorpholine | |
| 3-morpholin-4-ylpropan-1-ol |
Specifications
| 4441-30-9 | |
| 240°C to 241°C | |
| C7H15NO2 | |
| MFCD01713533 | |
| 4-3-hydroxypropyl morpholine, 3-morpholinopropanol, 4-morpholinepropanol, 3-morpholin-4-yl propan-1-ol, 3-morpholinopropan-1-ol, 3-4-morpholinyl-1-propanol, 3-morpholin-4-yl-propan-1-ol, 3-morpholino-1-propanol, gamma-morpholinopropanol german, n-hydroxypropanylmorpholine | |
| C1COCCN1CCCO | |
| 145.202 | |
| 145.2 | |
| 3-(4-Morpholinyl)-1-propanol |
| 1.049 | |
| 85°C (185°F) | |
| 1.477 | |
| 1 g | |
| VZKSLWJLGAGPIU-UHFFFAOYSA-N | |
| 3-morpholin-4-ylpropan-1-ol | |
| 199576 | |
| 95% |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H335
RTECSNumber : QE9104700
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only