missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)-1-propanol, 95%

Catalog No. AAH6037603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6037603 1 g
AAH6037606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6037603 Supplier Thermo Scientific Chemicals Supplier No. H6037603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4441-30-9 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4441-30-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD01713533
InChI Key VZKSLWJLGAGPIU-UHFFFAOYSA-N
Synonym 4-3-hydroxypropyl morpholine, 3-morpholinopropanol, 4-morpholinepropanol, 3-morpholin-4-yl propan-1-ol, 3-morpholinopropan-1-ol, 3-4-morpholinyl-1-propanol, 3-morpholin-4-yl-propan-1-ol, 3-morpholino-1-propanol, gamma-morpholinopropanol german, n-hydroxypropanylmorpholine
PubChem CID 199576
IUPAC Name 3-morpholin-4-ylpropan-1-ol
SMILES C1COCCN1CCCO

Specifications

Density 1.049
Boiling Point 240°C to 241°C
Flash Point 85°C (185°F)
Refractive Index 1.477
Quantity 1 g
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 3-(4-Morpholinyl)-1-propanol
Hazard Category H227-H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
RTECSNumber QE9104700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only