missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, ≥95%, Thermo Scientific™

Chemical Identifiers

CAS 859850-88-7
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
MDL Number MFCD08271925
InChI Key KKYYCUZQPBFXKD-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
PubChem CID 18525774
IUPAC Name 3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
SMILES CN1CCN(CC1)CC2=CC=CC(=C2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC45304CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45304DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45304DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

859850-88-7
218.3
KKYYCUZQPBFXKD-UHFFFAOYSA-N
18525774
CN1CCN(CC1)CC2=CC=CC(=C2)C=O
C13H18N2O
MFCD08271925
3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde

Specifications

859850-88-7
100.0
Amber Glass Bottle
MFCD08271925
KKYYCUZQPBFXKD-UHFFFAOYSA-N
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
18525774
95%
94.5
95%
C13H18N2O
3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
CN1CCN(CC1)CC2=CC=CC(=C2)C=O
218.3
218.3
3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents