missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylpiperazin-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 215309-01-6
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD06742238
InChI Key HALATUFUWLWCQV-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
PubChem CID 4741681
IUPAC Name 3-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC35901CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC35901DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

215309-01-6
220.27
HALATUFUWLWCQV-UHFFFAOYSA-N
4741681
CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O
C12H16N2O2
MFCD06742238
3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
3-(4-methylpiperazin-1-yl)benzoic acid

Specifications

215309-01-6
100.0
C12H16N2O2
3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O
220.27
220.27
3-(4-Methylpiperazin-1-yl)benzoic acid
96.5
97%
MFCD06742238
HALATUFUWLWCQV-UHFFFAOYSA-N
3-(4-methylpiperazin-1-yl)benzoic acid
4741681
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents