missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxyphenylacetonitrile 98.0+%, TCI America™

Catalog No. p-7132914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19235G 5 g
M192325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19235G Supplier TCI America Supplier No. M19235G
Only null left

Chemical Identifiers

CAS 4439-02-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005835
InChI Key ZQPBOYASBNAXOZ-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy phenylacetonitrile, 1,3-benzodioxole-5-acetonitrile, 3,4-methylenedioxyphenylacetonitrile, 2-benzo d 1,3 dioxol-5-yl acetonitrile, 3,4-methylenedioxybenzyl cyanide, 2-2h-1,3-benzodioxol-5-yl acetonitrile, 1,3-benzodioxol-5-ylacetonitrile, 2-1,3-benzodioxol-5-yl acetonitrile, 3,4-methylenedioxybenzylcyanide, homopiperonylonitrile
PubChem CID 78178
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetonitrile
SMILES C1OC2=C(O1)C=C(C=C2)CC#N

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Methylenedioxyphenylacetonitrile
EINECSNumber (5)-3564
TSCA No