missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7132494
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10085G 5 g
M100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10085G Supplier TCI America Supplier No. M10085G
Only null left

Chemical Identifiers

CAS 2861-28-1
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00014576
InChI Key ODVLMCWNGKLROU-UHFFFAOYSA-N
Synonym 1,3-benzodioxole-5-acetic acid, 3,4-methylenedioxy phenylacetic acid, 2-benzo d 1,3 dioxol-5-yl acetic acid, 3,4-methylenedioxyphenylacetic acid, homopiperonylic acid, benzo-1,3-dioxole-5-acetic acid, 1,3-benzodioxol-5-yl acetic acid, 2-2h-1,3-benzodioxol-5-yl acetic acid, 2-1,3-benzodioxol-5-yl acetic acid, 1,3-benzodioxol-5-ylacetic acid
PubChem CID 76115
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetic acid
SMILES C1OC2=C(O1)C=C(C=C2)CC(=O)O

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Methylenedioxyphenylacetic Acid