missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Methylenedioxynitrobenzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 148°C |
| Color | Yellow |
| Quantity | 500 g |
| Formula Weight | 167.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Methylenedioxynitrobenzene |
Safety and Handling
| TSCA | Yes |