missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxynitrobenzene 98.0+%, TCI America™

Catalog No. M084825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M084825G 25 g
M0848500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M084825G Supplier TCI America Supplier No. M084825G
Only null left

Chemical Identifiers

CAS 2620-44-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00005824
InChI Key SNWQAKNKGGOVMO-UHFFFAOYSA-N
Synonym 5-nitrobenzo d 1,3 dioxole, 1,2-methylenedioxy-4-nitrobenzene, 3,4-methylenedioxynitrobenzene, 1,3-benzodioxole, 5-nitro, methylenedioxynitrobenzene, 5-nitrobenzodioxole, 3,4-methylenedioxy-1-nitrobenzene, 5-nitro-2h-1,3-benzodioxole, benzene, 1,2-methylenedioxy-4-nitro
PubChem CID 75798
IUPAC Name 5-nitro-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 148°C
Color Yellow
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Methylenedioxynitrobenzene