missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxyacetophenone, 98%

Catalog No. AC330810050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330810050 5 g Glass bottle
AC330810250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330810050 Supplier Thermo Scientific Chemicals Supplier No. 330810050

CAS: 3162-29-6 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3162-29-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005831
InChI Key BMHMKWXYXFBWMI-UHFFFAOYSA-N
Synonym 3',4'-methylenedioxy acetophenone, 1-benzo d 1,3 dioxol-5-yl ethanone, 5-acetyl-1,3-benzodioxole, 3,4-methylenedioxyacetophenone, 3,4-methylenedioxy acetophenone, 3',4'-methylenedioxyacetophenone, 1-1,3-benzodioxol-5-yl ethanone, acetopiperone, 1-1,3-benzodioxol-5-yl ethan-1-one, ethanone, 1-1,3-benzodioxol-5-yl
PubChem CID 76622
IUPAC Name 1-(1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCO2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 89°C
Color Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Methylenedioxyacetophenone, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-613-1
TSCA TSCA

RUO – Research Use Only