missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylglyoxal hydrate, 97%, dry wt basis

Catalog No. p-7048129
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027303 1 g
AAL2027306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2027303 Supplier Thermo Scientific Chemicals Supplier No. L2027303
Only null left

CAS: 65709-23-1 | C9H6O4 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65709-23-1
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.14
MDL Number MFCD03411518
InChI Key OUCBKIWZWGSDJL-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenylglyoxal hydrate, 2-benzo d 1,3 dioxol-5-yl-2-oxoacetaldehyde hydrate, 3,4-methylenedioxy phenylglyoxal hydrate, 2-2h-1,3-benzodioxol-5-yl-2-oxoacetaldehyde hydrate, pubchem12143, benzo d 1,3 dioxole-5-glyoxal hydrate, 2-1,3-benzodioxol-5-yl-2-oxoacetaldehyde hydrate, 2-1,3-benzodioxol-5-yl-2-oxidanylidene-ethanal hydrate, 2-2h-benzo 3,4-d 1,3-dioxolan-5-yl-2-oxoethanal, hydrate
PubChem CID 15608490
IUPAC Name 2-(1,3-benzodioxol-5-yl)-2-oxoacetaldehyde;hydrate
SMILES O=CC(=O)C1=CC2=C(OCO2)C=C1

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Beilstein 7703971
Formula Weight 178.15 (Anhydrous)
Percent Purity 97%
Assay (dry wt.)
Chemical Name or Material 3,4-(Methylenedioxy)phenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only